Search
Search

ABHD18| Gene Symbol: ABHD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097507.3 | XP_011952897.1 | 5 - 6 | 583 | 98 | |
| XM_012097508.3 | XP_011952898.1 | 5 - 6 | 650 | 98 | ||
| XM_012097510.3 | XP_011952900.1 | 2 - 3 | 175 | 98 | ||
| XM_027956282.1 | XP_027812083.1 | 5 - 6 | 584 | 98 | ||
| XM_027956283.1 | XP_027812084.1 | 5 - 6 | 583 | 98 | ||
Set Membership: |
|