Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021467.3 | XP_004021516.2 | 12 - 13 | 1462 | 65 | |
| XM_027962201.1 | XP_027818002.1 | 10 - 11 | 1584 | 65 | ||
| XM_027962202.1 | XP_027818003.1 | 10 - 11 | 1589 | 65 | ||
| XM_027962203.1 | XP_027818004.1 | 10 - 11 | 1593 | 65 | ||
| XM_027962204.1 | XP_027818005.1 | 10 - 11 | 1595 | 65 | ||
| XM_027962205.1 | XP_027818006.1 | 8 - 9 | 1025 | 65 | ||
Set Membership: |
|