Search
Search

SEC31B| Gene Symbol: SEC31B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012102790.3 | XP_011958180.2 | 11 - 12 | 1278 | 54 | |
| XM_012102791.3 | XP_011958181.2 | 10 - 11 | 1269 | 54 | ||
| XM_012102792.3 | XP_011958182.2 | 10 - 11 | 1266 | 54 | ||
| XM_027960180.1 | XP_027815981.1 | 9 - 10 | 1071 | 54 | ||
Set Membership: |
|