Search
Search
| Gene Symbol: CSN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009378.1 | NP_001009378.1 | 2 - 3 | 129 | 91 | |
| GenBank mRNA | KC963133.1 | - | 2 - 3 | 106 | 91 | |
| KC963134.1 | - | 2 - 3 | 106 | 91 | ||
| X51822.1 | - | 129 | 91 | |||
Set Membership: |
|