Search
Search

CAPN3| Gene Symbol: CAPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009212.1 | NP_001009212.1 | 20 - 21 | 2233 | 85 | |
| GenBank mRNA | AF087570.1 | - | 20 - 21 | 2233 | 85 | |
| EU846598.1 | - | 17 - 18 | 1849 | 85 | ||
| KM250080.1 | - | 20 - 21 | 2168 | 85 | ||
| KM250081.1 | - | 20 - 21 | 2168 | 85 | ||
| KM250082.1 | - | 20 - 21 | 2169 | 85 | ||
Set Membership: |
|