Search
Search

SLC25A14| Gene Symbol: SLC25A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256816.1 | NP_001243745.1 | 2 - 3 | 311 | 71 | |
| XM_012106124.3 | XP_011961514.2 | 2 - 3 | 158 | 71 | ||
| XM_027962759.1 | XP_027818560.1 | 3 - 4 | 414 | 71 | ||
| XM_027962760.1 | XP_027818561.1 | 3 - 4 | 405 | 71 | ||
| XM_027962761.1 | XP_027818562.1 | 3 - 4 | 343 | 71 | ||
| GenBank mRNA | JQ278709.1 | - | 2 - 3 | 311 | 71 | |
Set Membership: |
|