Search
Search

VDAC1
LOC114116592| Gene Symbol: VDAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126352.1 | NP_001119824.1 | 1 | 547 | 106 | |
| XM_015095469.2 | XP_014950955.1 | 6 - 7 | 660 | 106 | ||
| XM_015095470.2 | XP_014950956.1 | 6 - 7 | 641 | 106 | ||
| XM_027969158.1 | XP_027824959.1 | 6 - 7 | 633 | 106 | ||
| GenBank mRNA | EU596596.1 | - | 1 | 547 | 106 | |
| Gene Symbol: LOC114116592 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973508.1 | XP_027829309.1 | 1 | 572 | 106 | |
Set Membership: |
|