Search
Search
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009279.1 | NP_001009279.1 | 3 - 4 | 624 | 61 | |
| XM_012103242.3 | XP_011958632.1 | 3 - 4 | 822 | 61 | ||
| XM_012103243.3 | XP_011958633.1 | 3 - 4 | 881 | 61 | ||
| XM_012103244.3 | XP_011958634.1 | 3 - 4 | 792 | 61 | ||
| GenBank mRNA | AY177612.1 | - | 3 - 4 | 615 | 61 | |
| AY206465.1 | - | 2 - 3 | 543 | 61 | ||
| AY304480.1 | - | 3 - 4 | 624 | 61 | ||
Set Membership: |
|