Search
Search

FLT3LG| Gene Symbol: FLT3LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078660.1 | NP_001072128.1 | 6 - 7 | 671 | 70 | |
| XM_027977374.1 | XP_027833175.1 | 7 - 8 | 817 | 70 | ||
| XM_027977375.1 | XP_027833176.1 | 7 - 8 | 843 | 70 | ||
| XM_027977376.1 | XP_027833177.1 | 7 - 8 | 763 | 70 | ||
| GenBank mRNA | DQ496115.1 | - | 6 - 7 | 671 | 70 | |
Set Membership: |
|