Search
Search

SCLT1| Gene Symbol: SCLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097481.2 | XP_011952871.2 | 9 - 10 | 1139 | 63 | |
| XM_012097484.2 | XP_011952874.1 | 5 - 6 | 421 | 63 | ||
| XM_027956277.1 | XP_027812078.1 | 9 - 10 | 1140 | 63 | ||
Set Membership: |
|