Search
Search

PPP3CA| Gene Symbol: PPP3CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970856.1 | XP_027826657.1 | 7 - 8 | 1632 | 67 | |
| XM_027970857.1 | XP_027826658.1 | 7 - 8 | 1633 | 67 | ||
Set Membership: |
|