Search
Search

DNAJB14| Gene Symbol: DNAJB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009673.4 | XP_004009722.1 | 5 - 6 | 802 | 76 | |
| XM_012179621.2 | XP_012035011.1 | 4 - 5 | 525 | 76 | ||
Set Membership: |
|