Search
Search

CASP8| Gene Symbol: CASP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012142477.3 | XP_011997867.1 | 8 - 9 | 1016 | 83 | |
| XM_012142484.3 | XP_011997874.1 | 7 - 8 | 972 | 83 | ||
| XM_012142493.3 | XP_011997883.1 | 6 - 7 | 909 | 83 | ||
Set Membership: |
|