Search
Search
| Gene Symbol: DAW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004986.4 | XP_004005035.3 | 10 - 11 | 1012 | 61 | |
| XM_027965269.1 | XP_027821070.1 | 10 - 11 | 953 | 61 | ||
| XM_027965270.1 | XP_027821071.1 | 9 - 10 | 886 | 61 | ||
Set Membership: |
|