Search
Search

OSBPL8| Gene Symbol: OSBPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006205.4 | XP_004006254.2 | 9 - 10 | 1184 | 61 | |
| XM_004006206.4 | XP_004006255.2 | 7 - 8 | 750 | 61 | ||
| XM_012173955.3 | XP_012029345.2 | 6 - 7 | 753 | 61 | ||
| XM_012173956.2 | XP_012029346.2 | 7 - 8 | 760 | 61 | ||
| XM_012173957.3 | XP_012029347.2 | 8 - 9 | 804 | 61 | ||
| XM_012173958.3 | XP_012029348.2 | 8 - 9 | 795 | 61 | ||
| XM_027967157.1 | XP_027822958.1 | 9 - 10 | 879 | 61 | ||
Set Membership: |
|