Search
Search

TMEM59| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002003.4 | XP_004002052.1 | 4 - 5 | 754 | 70 | |
| XM_012149467.3 | XP_012004857.1 | 5 - 6 | 656 | 70 | ||
| XM_012149537.3 | XP_012004927.1 | 5 - 6 | 636 | 70 | ||
| XM_012149705.3 | XP_012005095.1 | 4 - 5 | 708 | 70 | ||
Set Membership: |
|