Search
Search
| Gene Symbol: TKFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959918.1 | XP_027815719.1 | 10 - 11 | 1074 | 71 | |
| XM_027959919.1 | XP_027815720.1 | 10 - 11 | 1286 | 71 | ||
| XM_027959920.1 | XP_027815721.1 | 9 - 10 | 884 | 71 | ||
Set Membership: |
|