Search
Search

TRAPPC12| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964035.1 | XP_027819836.1 | 2 - 3 | 1319 | 62 | |
| XM_027964036.1 | XP_027819837.1 | 3 - 4 | 1824 | 62 | ||
| XM_027964037.1 | XP_027819838.1 | 3 - 4 | 1821 | 62 | ||
| XM_027964038.1 | XP_027819839.1 | 2 - 3 | 1715 | 62 | ||
| XM_027964039.1 | XP_027819840.1 | 2 - 3 | 1264 | 62 | ||
Set Membership: |
|