Search
Search

ZDHHC5| Gene Symbol: ZDHHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979801.1 | XP_027835602.1 | 6 - 7 | 1826 | 71 | |
| XM_027979802.1 | XP_027835603.1 | 6 - 7 | 743 | 71 | ||
Set Membership: |
|