Search
Search

IKBKB| Gene Symbol: IKBKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962683.1 | XP_027818484.1 | 12 - 13 | 1344 | 64 | |
| XM_027962684.1 | XP_027818485.1 | 7 - 8 | 1017 | 64 | ||
| XM_027962685.1 | XP_027818486.1 | 12 - 13 | 1314 | 64 | ||
Set Membership: |
|