Search
Search

PRSS42| Gene Symbol: PRSS42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957976.1 | XP_027813777.1 | 3 - 4 | 694 | 96 | |
| XM_027957977.1 | XP_027813778.1 | 4 - 5 | 784 | 96 | ||
Set Membership: |
|