Search
Search

SYTL2| Gene Symbol: SYTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019404.4 | XP_004019453.1 | 17 - 18 | 2886 | 73 | |
| XM_012101639.3 | XP_011957029.1 | 16 - 17 | 2838 | 73 | ||
| XM_012101640.3 | XP_011957030.1 | 16 - 17 | 2766 | 73 | ||
| XM_012101642.3 | XP_011957032.1 | 8 - 9 | 2107 | 73 | ||
| XM_027959800.1 | XP_027815601.1 | 10 - 11 | 3280 | 73 | ||
| XM_027959801.1 | XP_027815602.1 | 9 - 10 | 3232 | 73 | ||
| XM_027959802.1 | XP_027815603.1 | 9 - 10 | 3159 | 73 | ||
| XM_027959803.1 | XP_027815604.1 | 18 - 19 | 3123 | 73 | ||
| XM_027959804.1 | XP_027815605.1 | 16 - 17 | 2325 | 73 | ||
Set Membership: |
|