Search
Search

CCDC88A| Gene Symbol: CCDC88A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254391.2 | XP_008252613.1 | 29 - 30 | 5691 | 88 | |
| XM_008254392.2 | XP_008252614.1 | 29 - 30 | 5687 | 88 | ||
| XM_008254395.2 | XP_008252617.1 | 28 - 29 | 5604 | 88 | ||
| XM_017340761.1 | XP_017196250.1 | 28 - 29 | 5607 | 88 | ||
| XM_017340762.1 | XP_017196251.1 | 27 - 28 | 5406 | 88 | ||
| XM_017340763.1 | XP_017196252.1 | 27 - 28 | 5403 | 88 | ||
Set Membership: |
|