Search
Search

OVGP1| Gene Symbol: OVGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082105.1 | NP_001075574.1 | 10 - 11 | 1171 | 103 | |
| GenBank mRNA | AF347052.3 | - | 10 - 11 | 1171 | 103 | |
Set Membership: |
|