Search
Search

PHKA1| Gene Symbol: PHKA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165917.1 | NP_001159389.1 | 5 - 6 | 982 | 100 | |
| XM_008272695.2 | XP_008270917.1 | 5 - 6 | 1008 | 100 | ||
| XM_017349631.1 | XP_017205120.1 | 5 - 6 | 1009 | 100 | ||
| GenBank mRNA | J03247.1 | - | 5 - 6 | 928 | 100 | |
| M64656.1 | - | 5 - 6 | 982 | 100 | ||
Set Membership: |
|