Search
Search

CELF3| Gene Symbol: CELF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171308.1 | NP_001164779.1 | 5 - 6 | 489 | 79 | |
| XM_008264045.2 | XP_008262267.1 | 5 - 6 | 1407 | 79 | ||
| XM_017345516.1 | XP_017201005.1 | 5 - 6 | 1404 | 79 | ||
| XM_017345517.1 | XP_017201006.1 | 5 - 6 | 1407 | 79 | ||
Set Membership: |
|