Search
Search

FAM228A| Gene Symbol: FAM228A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254801.2 | XP_008253023.1 | 4 - 5 | 584 | 94 | |
| XM_008254802.2 | XP_008253024.1 | 3 - 4 | 392 | 94 | ||
Set Membership: |
|