Search
Search

HEATR9| Gene Symbol: HEATR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271141.2 | XP_008269363.2 | 9 - 10 | 1335 | 69 | |
| XM_017349036.1 | XP_017204525.1 | 9 - 10 | 1337 | 69 | ||
Set Membership: |
|