Search
Search

LARGE2| Gene Symbol: LARGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709074.3 | XP_002709120.1 | 3 - 4 | 537 | 93 | |
| XM_008270235.2 | XP_008268457.1 | 3 - 4 | 526 | 93 | ||
| XM_008270241.2 | XP_008268463.1 | 4 - 5 | 458 | 93 | ||
| XM_008270253.2 | XP_008268475.1 | 4 - 5 | 458 | 93 | ||
Set Membership: |
|