Search
Search

LOXL3| Gene Symbol: LOXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254204.2 | XP_008252426.1 | 16 - 17 | 2727 | 58 | |
| XM_008254205.2 | XP_008252427.1 | 15 - 16 | 2668 | 58 | ||
| XM_008254206.2 | XP_008252428.1 | 14 - 15 | 2545 | 58 | ||
| XM_008254207.2 | XP_008252429.1 | 16 - 17 | 2404 | 58 | ||
| XM_008254209.2 | XP_008252431.1 | 16 - 17 | 2398 | 58 | ||
| XM_008254210.2 | XP_008252432.1 | 10 - 11 | 1462 | 58 | ||
Set Membership: |
|