Search
Search

RNF167| Gene Symbol: RNF167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718831.3 | XP_002718877.1 | 7 - 8 | 1097 | 64 | |
| XM_017348861.1 | XP_017204350.1 | 6 - 7 | 1122 | 64 | ||
| XM_017348862.1 | XP_017204351.1 | 6 - 7 | 541 | 64 | ||
Set Membership: |
|