Search
Search

KDM5C| Gene Symbol: KDM5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720016.3 | XP_002720062.1 | 18 - 19 | 2618 | 94 | |
| XM_008272647.2 | XP_008270869.1 | 18 - 19 | 2618 | 94 | ||
| XM_008272648.2 | XP_008270870.1 | 18 - 19 | 2615 | 94 | ||
| XM_017349757.1 | XP_017205246.1 | 17 - 18 | 3024 | 94 | ||
| XM_017349758.1 | XP_017205247.1 | 17 - 18 | 2495 | 94 | ||
| XM_017349759.1 | XP_017205248.1 | 18 - 19 | 3147 | 94 | ||
Set Membership: |
|