Search
Search

ARHGEF25| Gene Symbol: ARHGEF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720920.3 | XP_002720966.1 | 1388 | 60 | ||
| XM_008274224.2 | XP_008272446.1 | 1358 | 60 | |||
| XM_017350486.1 | XP_017205975.1 | 2183 | 60 | |||
| XM_017350487.1 | XP_017205976.1 | 2167 | 60 | |||
| XM_017350488.1 | XP_017205977.1 | 1323 | 60 | |||
Set Membership: |
|