Search
Search

CNPY4| Gene Symbol: CNPY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248886.2 | XP_008247108.1 | 589 | 57 | ||
| XM_008248887.2 | XP_008247109.1 | 475 | 57 | |||
| XM_017337870.1 | XP_017193359.1 | 561 | 57 | |||
Set Membership: |
|