Search
Search

LOC100357938| Gene Symbol: LOC100357938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719508.3 | XP_002719554.2 | 4 - 5 | 1965 | 97 | |
| XM_008271127.2 | XP_008269349.1 | 4 - 5 | 2129 | 97 | ||
Set Membership: |
|