Search
Search

HVCN1| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272226.2 | XP_008270448.1 | 5 - 6 | 529 | 67 | |
| XM_008272227.2 | XP_008270449.1 | 6 - 7 | 568 | 67 | ||
| XM_008272231.2 | XP_008270453.1 | 4 - 5 | 489 | 67 | ||
| XM_017349524.1 | XP_017205013.1 | 5 - 6 | 530 | 67 | ||
Set Membership: |
|