Search
Search

SCCPDH| Gene Symbol: SCCPDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717629.3 | XP_002717675.1 | 1 - 2 | 314 | 118 | |
| XM_017347699.1 | XP_017203188.1 | 1 - 2 | 317 | 118 | ||
Set Membership: |
|