Search
Search

LRRC6| Gene Symbol: LRRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008255918.2 | XP_008254140.1 | 5 - 6 | 954 | 109 | |
| XM_017341540.1 | XP_017197029.1 | 5 - 6 | 954 | 109 | ||
Set Membership: |
|