Search
Search

LOC4337286| Gene Symbol: LOC4337286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777946.2 | XP_015633432.1 | 2 - 3 | 344 | 95 | |
| XM_015777947.2 | XP_015633433.1 | 2 - 3 | 337 | 95 | ||
| GenBank mRNA | AK102842.1 | - | 2 - 3 | 293 | 95 | |
Set Membership: |
|