Search
Search

LOC4340521| Gene Symbol: LOC4340521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788845.2 | XP_015644331.1 | 3 - 4 | 981 | 145 | |
| GenBank mRNA | AK102978.1 | - | 3 - 4 | 960 | 145 | |
Set Membership: |
|