Search
Search

LOC4327611| Gene Symbol: LOC4327611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765748.2 | XP_015621234.1 | 1 - 2 | 348 | 119 | |
| GenBank mRNA | AK108657.1 | - | 1 - 2 | 350 | 119 | |
| AY870606.1 | - | 1 - 2 | 341 | 119 | ||
Set Membership: |
|