Search
Search

LOC4338624| Gene Symbol: LOC4338624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785243.2 | XP_015640729.1 | 1 - 2 | 246 | 144 | |
| GenBank mRNA | AK069380.1 | - | 1 - 2 | 243 | 144 | |
| AK102118.1 | - | 1 - 2 | 247 | 144 | ||
Set Membership: |
|