Search
Search

LOC4331517| Gene Symbol: LOC4331517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773237.2 | XP_015628723.1 | 5 - 6 | 510 | 107 | |
| GenBank mRNA | AK065483.1 | - | 5 - 6 | 502 | 107 | |
Set Membership: |
|