Search
Search

LOC4334938| Gene Symbol: LOC4334938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778227.2 | XP_015633713.1 | 3 - 4 | 430 | 111 | |
| GenBank mRNA | AK073153.1 | - | 3 - 4 | 431 | 111 | |
| AK121833.1 | - | 3 - 4 | 388 | 111 | ||
Set Membership: |
|