Search
Search

LOC4344225| Gene Symbol: LOC4344225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790078.2 | XP_015645564.1 | 1 - 2 | 419 | 67 | |
| GenBank mRNA | AK061122.1 | - | 1 - 2 | 419 | 67 | |
Set Membership: |
|