Search
Search

LOC4327914| Gene Symbol: LOC4327914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771623.2 | XP_015627109.1 | 2 - 3 | 586 | 57 | |
| GenBank mRNA | AJ495786.1 | - | 2 - 3 | 536 | 57 | |
| AK111803.1 | - | 2 - 3 | 541 | 57 | ||
| FJ940218.1 | - | 2 - 3 | 519 | 57 | ||
Set Membership: |
|