Search
Search

LOC4341106| Gene Symbol: LOC4341106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788747.2 | XP_015644233.1 | 2 - 3 | 701 | 89 | |
| GenBank mRNA | AK099764.1 | - | 2 - 3 | 704 | 89 | |
| AK103794.1 | - | 2 - 3 | 643 | 89 | ||
| AK121899.1 | - | 2 - 3 | 643 | 89 | ||
Set Membership: |
|