Search
Search

LOC4344326| Gene Symbol: LOC4344326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792404.2 | XP_015647890.1 | 1 - 2 | 157 | 138 | |
| GenBank mRNA | AK065816.1 | - | 1 - 2 | 161 | 138 | |
| AK104095.1 | - | 1 - 2 | 153 | 138 | ||
| AK119625.1 | - | 1 - 2 | 161 | 138 | ||
Set Membership: |
|