Search
Search

LOC4342343| Gene Symbol: LOC4342343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791957.2 | XP_015647443.1 | 3 - 4 | 602 | 65 | |
| GenBank mRNA | AK103862.1 | - | 2 - 3 | 419 | 65 | |
| AK106110.1 | - | 3 - 4 | 545 | 65 | ||
| AK120257.1 | - | 3 - 4 | 547 | 65 | ||
| AY224447.1 | - | 351 | 65 | |||
Set Membership: |
|