Search
Search

GALNTL5| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135320.2 | XP_014990806.1 | 1531 | 70 | ||
| XM_015135321.2 | XP_014990807.1 | 1412 | 70 | |||
| XM_015135324.2 | XP_014990810.1 | 1239 | 70 | |||
| XM_015135325.2 | XP_014990811.1 | 1120 | 70 | |||
| XM_028846495.1 | XP_028702328.1 | 1241 | 70 | |||
| XM_028846496.1 | XP_028702329.1 | 1097 | 70 | |||
Set Membership: |
|